Thermo Scientific Chemicals

2-Ethyl-1-butene, 97%, Thermo Scientific Chemicals

Catalog number: L11553.14
25 g, Each
Thermo Scientific Chemicals

2-Ethyl-1-butene, 97%, Thermo Scientific Chemicals

Catalog number: L11553.14
25 g, Each
Quantity
Catalog number: L11553.14
also known as L11553-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
760-21-4
IUPAC Name
3-methylidenepentane
Molecular Formula
C6H12
InChI Key
RYKZRKKEYSRDNF-UHFFFAOYSA-N
SMILES
CCC(=C)CC
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Form
Liquid
Refractive Index
1.3945-1.3995 @ 20?C
Assay (GC)
≥96.0%

Description

2-Ethyl-1-butene is used as an intermediate in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyl-1-butene is used as an intermediate in organic synthesis and pharmaceuticals.

Solubility
Soluble in alcohol, and methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text