Thermo Scientific Chemicals

Propargyl methacrylate, 98%, stab. with 200-300ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L11556.09
10 g, Each
Thermo Scientific Chemicals

Propargyl methacrylate, 98%, stab. with 200-300ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L11556.09
10 g, Each
Quantity
Catalog number: L11556.09
also known as L11556-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13861-22-8
IUPAC Name
prop-2-yn-1-yl 2-methylprop-2-enoate
Molecular Formula
C7H8O2
InChI Key
PZAWASVJOPLHCJ-UHFFFAOYSA-N
SMILES
CC(=C)C(=O)OCC#C
Form
Liquid
Assay (GC)
>97.5%
Appearance (Color)
Clear, almost colorless
Stabilizer
200-300ppm 4-methoxyphenol

Description

Propargyl methacrylate is a monomer used in polymerization reactions. It is associated with its acetylene and under goes polymerization reaction through the reversible addition fragmentation chain transfer (RAFT) process using cyanoisopropyl dithiobenzoate as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propargyl methacrylate is a monomer used in polymerization reactions. It is associated with its acetylene and under goes polymerization reaction through the reversible addition fragmentation chain transfer (RAFT) process using cyanoisopropyl dithiobenzoate as a reagent.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text