3,3'-Dimethoxybenzil, 99+%, Thermo Scientific Chemicals
3,3'-Dimethoxybenzil, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3'-Dimethoxybenzil, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L11564.03
also known as L11564-03
Price (USD)/ Each
36.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
36.50
Add to cart
3,3'-Dimethoxybenzil, 99+%, Thermo Scientific Chemicals
Catalog numberL11564.03
Price (USD)/ Each
36.50
-
Add to cart
Chemical Identifiers
CAS40101-17-5
IUPAC Namebis(3-methoxyphenyl)ethane-1,2-dione
Molecular FormulaC16H14O4
InChI KeyPJGXOGKIVAJFTE-UHFFFAOYSA-N
SMILESCOC1=CC(=CC=C1)C(=O)C(=O)C1=CC(OC)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
Assay (GC)≥99.0%
Melting Point (clear melt)79.0-85.0°C
Identification (FTIR)Conforms (non-U.S. specification)
FormCrystals or powder or crystalline powder or granules
View more
3,3'-Dimethoxybenzil used in the preparation of benzil derivatives. It is also used as an intermediate in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′-Dimethoxybenzil used in the preparation of benzil derivatives. It is also used as an intermediate in organic synthesis and pharmaceuticals.

Solubility
Insoluble in water.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ergang Wang.; Jonas Bergqvist.; Koen Vandewal.; Zaifei Ma.; Lintao Hou.; Angelica Lundin.; Scott Himmelberger.; Alberto Salleo.; Christian Müller.; Olle Inganäs.;Fengling Zhang.; Mats R. Andersson. Conformational Disorder Enhances Solubility and Photovoltaic Performance of a Thiophene-Quinoxaline Copolymer. Advanced Energy Materials. 2013, 2 (6), 806-814.
  2. Darren J. Goossens.; T. Richard Welberry.; Aidan P. Heerdegen.; Alison J. Edwards. Modelling disorder in 3,3'-dimethoxybenzil, C16H14O4. Zeitschrift für Kristallographie. 2005, 220 (12), 1035-1042.