Thermo Scientific Chemicals

3,3'-Dimethoxybenzil, 99+%, Thermo Scientific Chemicals

Catalog number: L11564.06
5 g, Each
Thermo Scientific Chemicals

3,3'-Dimethoxybenzil, 99+%, Thermo Scientific Chemicals

Catalog number: L11564.06
5 g, Each
Quantity
Catalog number: L11564.06
also known as L11564-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
40101-17-5
IUPAC Name
bis(3-methoxyphenyl)ethane-1,2-dione
Molecular Formula
C16H14O4
InChI Key
PJGXOGKIVAJFTE-UHFFFAOYSA-N
SMILES
COC1=CC(=CC=C1)C(=O)C(=O)C1=CC(OC)=CC=C1
Appearance (Color)
Pale yellow to yellow
Assay (GC)
≥99.0%
Melting Point (clear melt)
79.0-85.0°C
Identification (FTIR)
Conforms (non-U.S. specification)
Form
Crystals or powder or crystalline powder or granules

Description

3,3'-Dimethoxybenzil used in the preparation of benzil derivatives. It is also used as an intermediate in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′-Dimethoxybenzil used in the preparation of benzil derivatives. It is also used as an intermediate in organic synthesis and pharmaceuticals.

Solubility
Insoluble in water.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text