Thermo Scientific Chemicals

5-Phenyl-1-pentyne, 98+%, Thermo Scientific Chemicals

Catalog number: L11608.06
5 g, Each
Thermo Scientific Chemicals

5-Phenyl-1-pentyne, 98+%, Thermo Scientific Chemicals

Catalog number: L11608.06
5 g, Each
Quantity
Catalog number: L11608.06
also known as L11608-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1823-14-9
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Refractive Index
1.5145-1.5185 @ 20?C (UK Sourced Material)
Form
Liquid or viscous liquid

Description

5-Phenyl-1-pentyne is also used as an intermediate in organic synthesis and pharmaceuticals. It is also used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Phenyl-1-pentyne is also used as an intermediate in organic synthesis and pharmaceuticals. It is also used to make other chemicals.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text