5-Phenyl-1-pentyne is also used as an intermediate in organic synthesis and pharmaceuticals. It is also used to make other chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Phenyl-1-pentyne is also used as an intermediate in organic synthesis and pharmaceuticals. It is also used to make other chemicals.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
- Elizabeth S. Roberts.; William L. Alworth.; Paul F. Hollenberga. Mechanism-Based Inactivation of Cytochromes P450 2E1 and 2B1 by 5-Phenyl-1-pentyne. Archives of Biochemistry and Biophysics. 1998, 354 (2), 295-302.
- Talitha M. Selby.; Timothy S. Zwier. Conformation-Specific Spectroscopy of 4-Phenyl-1-butyne and 5-Phenyl-1-pentyne. J. Phys. Chem. A. 2005, 109 (38), 8487-8496.