Thermo Scientific Chemicals

N-Phenyl-o-phenylenediamine, 98%, Thermo Scientific Chemicals

Catalog number: L11616.06
5 g, Each
Thermo Scientific Chemicals

N-Phenyl-o-phenylenediamine, 98%, Thermo Scientific Chemicals

Catalog number: L11616.06
5 g, Each
Quantity
Catalog number: L11616.06
also known as L11616-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
534-85-0
Appearance (Color)
Red to dark brown to brown or pink
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder or fused solid
Assay (GC)
≥97.5%

Description

N-Phenyl-o-phenylenediamine is also used as an intermediate in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Phenyl-o-phenylenediamine is also used as an intermediate in organic synthesis and pharmaceuticals.

Solubility
Soluble in acetone, chloroform and benzene. Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to light. Store at room temperature. Incompatible with acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text