N-Phenyl-o-phenylenediamine is also used as an intermediate in organic synthesis and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Phenyl-o-phenylenediamine is also used as an intermediate in organic synthesis and pharmaceuticals.
Solubility
Soluble in acetone, chloroform and benzene. Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to light. Store at room temperature. Incompatible with acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
RUO – Research Use Only
General References:
- Maslivets, A. N.; L. I. Smirnova.; Yu S. Andreichikov. 5-Membered 2, 3-Dioxoheterocycles. 31. Interaction Of 1-Aryl-4-Aroyl-5-Methoxycarbonyl-2, 3-Dihydro-2, 3-Pyrrolediones With O-Aminophenol And N-Phenyl-O-Phenylenediamine. Zhurnal Organicheskoi Khimii . 1992, 28 (10), 2141-2148.
- EVELYN CUEVAS CREENCIA.; TAGUCHI Kei.; HORAGUCHI Takaaki. Thermal Reactions of N-Alkyl-2-benzylaniline and N-Alkyl-N'-phenyl-o-phenylenediamine : An Unusual Route to 2-Phenylindole and 2-Phenylbenzimidazole. Journal of heterocyclic chemistry . 2008, 45 (3), 837-843.