Thermo Scientific Chemicals

Aminoguanidine nitrate, 98%, cont. ca 10% water, Thermo Scientific Chemicals

Catalog number: L11632.22
100 g, Each
Thermo Scientific Chemicals

Aminoguanidine nitrate, 98%, cont. ca 10% water, Thermo Scientific Chemicals

Catalog number: L11632.22
100 g, Each
Quantity
Catalog number: L11632.22
also known as L11632-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10308-82-4
IUPAC Name
N''-aminoguanidine; nitric acid
Molecular Formula
CH7N5O3
InChI Key
PMGFHEJUUBDCLU-UHFFFAOYSA-N
SMILES
O[N+]([O-])=O.NN=C(N)N
Appearance (Color)
White
Form
Crystals or Crystalline powder
Assay (Non-aqueous acid-base Titration)
>97.5 to <102.5% (dry wt. basis)
Water Content (Karl Fischer Titration)
<20%

Description

Aminoguanidine nitrate is used as a pharmaceutical raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aminoguanidine nitrate is used as a pharmaceutical raw material.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with reducing agents, strong oxidizing agents, organic materials and metal powders.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text