Thermo Scientific Chemicals

3-(Trifluoromethyl)phenyl isothiocyanate, 98%, Thermo Scientific Chemicals

Catalog number: L11646.06
5 g, Each
Thermo Scientific Chemicals

3-(Trifluoromethyl)phenyl isothiocyanate, 98%, Thermo Scientific Chemicals

Catalog number: L11646.06
5 g, Each
Quantity
Catalog number: L11646.06
also known as L11646-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1840-19-3
IUPAC Name
1-isothiocyanato-3-(trifluoromethyl)benzene
Molecular Formula
C8H4F3NS
InChI Key
GFEPANUKFYVALF-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC=CC(=C1)N=C=S
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%
Comment
Specification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)
Conforms (non-U.S. specification)

Description

Used as pharamaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharamaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text