4-Morpholinepropionitrile, 98+%, Thermo Scientific Chemicals
4-Morpholinepropionitrile, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Morpholinepropionitrile, 98+%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Catalog number L11664.14
also known as L11664-14
Price (USD)/ Each
92.10
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
92.10
Add to cart
4-Morpholinepropionitrile, 98+%, Thermo Scientific Chemicals
Catalog numberL11664.14
Price (USD)/ Each
92.10
-
Add to cart
Chemical Identifiers
CAS4542-47-6
IUPAC Name3-(morpholin-4-yl)propanenitrile
Molecular FormulaC7H12N2O
InChI KeyWXVKGHVDWWXBJX-UHFFFAOYSA-N
SMILESN#CCCN1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4685-1.4725 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only