FormCrystals or powder or crystalline powder or fused solid
Assay from Suppliers CofA≥98.5% (U.S. specification)
Optical Rotation28.0 ± 1.5° (C=1.5 in 1N NaOH, non-U.S. specification))
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5% (non-U.S. specification)
View more