5-Chloro-7-methylisatin, 97%, Thermo Scientific Chemicals
5-Chloro-7-methylisatin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Chloro-7-methylisatin, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number L11682.MD
also known as L11682-MD
Price (USD)/ Each
109.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
109.00
Add to cart
5-Chloro-7-methylisatin, 97%, Thermo Scientific Chemicals
Catalog numberL11682.MD
Price (USD)/ Each
109.00
-
Add to cart
Chemical Identifiers
CAS14389-06-1
IUPAC Name5-chloro-7-methyl-2,3-dihydro-1H-indole-2,3-dione
Molecular FormulaC9H6ClNO2
InChI KeyLDFQLYHDZZPAGN-UHFFFAOYSA-N
SMILESCC1=C2NC(=O)C(=O)C2=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
FormPowder
Appearance (Color)Orange to red to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only