Thermo Scientific Chemicals

3,5-Dimethylphenyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L11698.03
1 g, Each
Thermo Scientific Chemicals

3,5-Dimethylphenyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L11698.03
1 g, Each
Quantity
Catalog number: L11698.03
also known as L11698-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
54132-75-1
IUPAC Name
1-isocyanato-3,5-dimethylbenzene
Molecular Formula
C9H9NO
InChI Key
DZSGDHNHQAJZCO-UHFFFAOYSA-N
SMILES
CC1=CC(=CC(C)=C1)N=C=O
Form
Liquid
Assay from Supplier's CofA
≥96.0% (US-sourced material)
Appearance (Color)
Clear colorless to pale yellow
Comment
Material sourced in UK and US
Refractive Index
1.5260-1.5310 @ 20?C (UK-sourced material)

Description

3,5-Dimethylphenyl isocyanate is applications include linking with oligosaccharides and cyclodextrins for chiral chromatography stationary phases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylphenyl isocyanate is applications include linking with oligosaccharides and cyclodextrins for chiral chromatography stationary phases.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text