Thermo Scientific Chemicals

4-Fluorophenylacetone, 99%, Thermo Scientific Chemicals

Catalog number: L11720.06
5 g, Each
Thermo Scientific Chemicals

4-Fluorophenylacetone, 99%, Thermo Scientific Chemicals

Catalog number: L11720.06
5 g, Each
Quantity
Catalog number: L11720.06
also known as L11720-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
459-03-0
IUPAC Name
1-(4-fluorophenyl)propan-2-one
Molecular Formula
C9H9FO
InChI Key
ZUEKIIWSVFBTCM-UHFFFAOYSA-N
SMILES
CC(=O)CC1=CC=C(F)C=C1
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥98.5%
Form
Liquid
Refractive Index
1.4945-1.4975 @ 20?C

Description

4-Fluorophenylacetone is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenylacetone is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text