Thermo Scientific Chemicals

1-Chloro-3,3-dimethylbutane, 97%, Thermo Scientific Chemicals

Catalog number: L11753.18
50 g, Each
Thermo Scientific Chemicals

1-Chloro-3,3-dimethylbutane, 97%, Thermo Scientific Chemicals

Catalog number: L11753.18
50 g, Each
Quantity
Catalog number: L11753.18
also known as L11753-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2855-08-5
IUPAC Name
1-chloro-3,3-dimethylbutane
Molecular Formula
C6H13Cl
InChI Key
XGCKOSFYXBAPQM-UHFFFAOYSA-N
SMILES
CC(C)(C)CCCl
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0% (non-U.S. specification)
Assay by NMR
≥96.0% (U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated

Description

1-Chloro-3,3-dimethylbutane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-3,3-dimethylbutane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Not miscible or difficult to mix in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text