trans-5-Decen-1-ol, 96%, Thermo Scientific Chemicals
trans-5-Decen-1-ol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-5-Decen-1-ol, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L11768.03
also known as L11768-03
Price (USD)/ Each
59.10
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
59.10
Add to cart
trans-5-Decen-1-ol, 96%, Thermo Scientific Chemicals
Catalog numberL11768.03
Price (USD)/ Each
59.10
-
Add to cart
Chemical Identifiers
CAS56578-18-8
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4480-1.4540 @ 20°C
FormLiquid or viscous liquid
Appearance (Color)Clear colorless to yellow
Assay (GC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only