Thermo Scientific Chemicals

10-Undecyn-1-ol, 96%, Thermo Scientific Chemicals

Catalog number: L11807.03
1 g, Each
Thermo Scientific Chemicals

10-Undecyn-1-ol, 96%, Thermo Scientific Chemicals

Catalog number: L11807.03
1 g, Each
Quantity
Catalog number: L11807.03
also known as L11807-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2774-84-7
IUPAC Name
undec-10-yn-1-ol
Molecular Formula
C11H20O
InChI Key
YUQZOUNRPZBQJK-UHFFFAOYSA-N
SMILES
OCCCCCCCCCC#C
Refractive Index
1.4535-1.4605 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid
Assay (GC)
≥95.0%
Appearance (Color)
Clear colorless to white to yellow

Description

10-Undecyn-1-ol exhibits an antifungal activity. It is also used in the esterification of pentanoic and stearic acids in the presence of lipase. It is also employed as a reagent in the preparation of 11-bromo-undec-1-yne by reacting with carbon tetrabromide and triphenylphosphine. Further, it acts as an intermediate in synthetic chemistry. In addition to this, it serves as a precursor in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
10-Undecyn-1-ol exhibits an antifungal activity. It is also used in the esterification of pentanoic and stearic acids in the presence of lipase. It is also employed as a reagent in the preparation of 11-bromo-undec-1-yne by reacting with carbon tetrabromide and triphenylphosphine. Further, it acts as an intermediate in synthetic chemistry. In addition to this, it serves as a precursor in organic synthesis.

Solubility
Immiscible with water.

Notes
Air sensitive. Incompatible strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text