Thermo Scientific Chemicals

N-Acetyl-L-alanine, 96%

Catalog number: L11811.03
1 g, Each
Thermo Scientific Chemicals

N-Acetyl-L-alanine, 96%

Catalog number: L11811.03
1 g, Each
Quantity
Catalog number: L11811.03
also known as L11811-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
97-69-8
IUPAC Name
2-acetamidopropanoic acid
Molecular Formula
C5H9NO3
InChI Key
KTHDTJVBEPMMGL-UHFFFAOYNA-N
SMILES
CC(NC(C)=O)C(O)=O
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%
Appearance (Color)
White
Optical Rotation
-65.5 ± 1.5? (c=2 in water)
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text