Thermo Scientific Chemicals

Phenyl acrylate, 97%, Thermo Scientific Chemicals

Catalog number: L11819.14
25 g, Each
Thermo Scientific Chemicals

Phenyl acrylate, 97%, Thermo Scientific Chemicals

Catalog number: L11819.14
25 g, Each
Quantity
Catalog number: L11819.14
also known as L11819-14
Price (USD)
Quantity
-
Have Questions?

Description

Phenyl acrylate reacts with benzaldehyde to produce 2-(hydroxy-phenyl-methyl)-acrylic acid phenyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl acrylate reacts with benzaldehyde to produce 2-(hydroxy-phenyl-methyl)-acrylic acid phenyl ester.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store at 4°C. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text