Ethylenediaminetetraacetic acid disodium magnesium salt hydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is used as chelating agents over a broad pH range in aqueous systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is used as chelating agents over a broad pH range in aqueous systems.
Solubility
Soluble in water. Insoluble in acid and organic liquids
Notes
Incompatible with oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Store at room temperature.
RUO – Research Use Only
W. J. Blaedel.; H. T. Knight. Purification and Properties of Disodium Salt of Ethylenediaminetetraacetic Acid as Primary Standard. Anal. Chem. 1954, 26 (4), 741-743.
Albert Darbey. Colorimetric Determination of Sodium Salts of Ethylenediaminetetraacetic Acid. Anal. Chem. 1952, 24 (2), 373-378.