Thermo Scientific Chemicals

Carbon powder, activated, Norit GSX, steam activated, acid washed, Thermo Scientific Chemicals

Catalog number: L11860.0P
2000 g, Each
Thermo Scientific Chemicals

Carbon powder, activated, Norit GSX, steam activated, acid washed, Thermo Scientific Chemicals

Catalog number: L11860.0P
2000 g, Each
Quantity
Catalog number: L11860.0P
also known as L11860-0P
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-44-0
IUPAC Name
carbon
Molecular Formula
C
InChI Key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES
[C]
Form
Powder
Loss on Drying
<10%
Appearance (Color)
Black
Comment
Molasses number ≤ 275

Description

Decolorizing carbon. High purity grade with high internal surface area, suitable for decolorization, purification and catalyst carrier applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text