Thermo Scientific Chemicals

4-Vinylbiphenyl, 97%, Thermo Scientific Chemicals

Catalog number: L11869.03
1 g, Each
Thermo Scientific Chemicals

4-Vinylbiphenyl, 97%, Thermo Scientific Chemicals

Catalog number: L11869.03
1 g, Each
Quantity
Catalog number: L11869.03
also known as L11869-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2350-89-2
Appearance (Color)
White to yellow to brown
Identification (FTIR)
Conforms
Melting Point (clear melt)
115-124?C
Solution Test
(1% in diethyl ether) : Clear
Assay (GC)
≥96.0%

Description

4-Vinylbiphenyl intermediate is used in organic synthesis and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Vinylbiphenyl intermediate is used in organic synthesis and in chemical research.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilat
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text