N,N-Dimethylglycine methyl ester is used as a biochemical for proteomics research. It is also used as chemical and pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N-Dimethylglycine methyl ester is used as a biochemical for proteomics research. It is also used as chemical and pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
It is sensitive to air and hygroscopic in nature. Protect from humidity and water. Store under dry inert gas. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
A. Gómez-Zavaglia.; R. Fausto. Low temperature FT-IR and molecular orbital study of N,N-dimethylglycine methyl ester: Proof for different ground conformational states in gas phase and in condensed media. Phys. Chem. Chem. Phys. 2003, 5 (1), 52-63.
Janusz Rak.; Piotr Skurski.; Maciej Gutowski. An ab initio study of the betaine anion-dipole-bound anionic state of a model zwitterion system. J. Chem. Phys. 2001, 114 (24), 10673.