Thermo Scientific Chemicals

N,N-Dimethylglycine methyl ester, 98%, Thermo Scientific Chemicals

Catalog number: L11897.14
25 g, Each
Thermo Scientific Chemicals

N,N-Dimethylglycine methyl ester, 98%, Thermo Scientific Chemicals

Catalog number: L11897.14
25 g, Each
Quantity
Catalog number: L11897.14
also known as L11897-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7148-06-3
IUPAC Name
methyl 2-(dimethylamino)acetate
Molecular Formula
C5H11NO2
InChI Key
LRZFEBJUJIQVDQ-UHFFFAOYSA-N
SMILES
COC(=O)CN(C)C
Form
liquid
Assay (GC)
>97.5%
Refractive Index
1.4105-1.4145
Appearance (Color)
Clear, colourless to pale yellow

Description

N,N-Dimethylglycine methyl ester is used as a biochemical for proteomics research. It is also used as chemical and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylglycine methyl ester is used as a biochemical for proteomics research. It is also used as chemical and pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
It is sensitive to air and hygroscopic in nature. Protect from humidity and water. Store under dry inert gas. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text