Thermo Scientific Chemicals

Thiazolyl Blue tetrazolium bromide, 98%

Catalog number: L11939.06
5 g, Each
Thermo Scientific Chemicals

Thiazolyl Blue tetrazolium bromide, 98%

Catalog number: L11939.06
5 g, Each
Quantity
Catalog number: L11939.06
also known as L11939-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
298-93-1
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Assay (Argentometric Titration)
≥97.5 to ≤102.5%
Appearance (Color)
White to yellow

Description

Used in measurement of cell proliferation. Produces a yellowish solution that is converted to dark blue, water-insoluble MTT formazan by mitochondrial dehydrogenases of living cells.Thiazolyl Blue tetrazolium bromide is a dye used in measurement of cell proliferation. It is used as a direct indicator of cytotoxicity (such as for screening cancer drugs) and apoptosis. As an electron acceptor, it is utilized for studying nicotinamide adenine dinucleotide phosphate (NADP)-dependent dehydrogenases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in measurement of cell proliferation. Produces a yellowish solution that is converted to dark blue, water-insoluble MTT formazan by mitochondrial dehydrogenases of living cells.Thiazolyl Blue tetrazolium bromide is a dye used in measurement of cell proliferation. It is used as a direct indicator of cytotoxicity (such as for screening cancer drugs) and apoptosis. As an electron acceptor, it is utilized for studying nicotinamide adenine dinucleotide phosphate (NADP)-dependent dehydrogenases.

Solubility
Soluble in water, ethanol, 2-methoxyethanol and phosphate buffered saline.

Notes
Light, air and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text