mono-n-Dodecyl phosphate is a lysophosphatidic acid (LPA) receptor ligand which serves as a specific agonist at EDG-4 (LPA2) (EC50=700 nM) and antagonist at EDG-7
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
mono-n-Dodecyl phosphate is a lysophosphatidic acid (LPA) receptor ligand which serves as a specific agonist at EDG-4 (LPA2) (EC50=700 nM) and antagonist at EDG-7
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
RUO – Research Use Only
General References:
- Arthur K. Nelson.; A. D. F. Toy. The Preparation of Long-Chain Monoalkyl Phosphates from Pyrophosphoric Acid and Alcohols. Inorg. Chem. 1963, 2 (4), 775-777.
- Stephanie M. Schmidt.; James McDonald.; Emma T. Pineda.; Ann M. Verwilst.; Yimei Chen.; Robert Josephs.; Agnes. E. Ostafin. Surfactant based assembly of mesoporous patterned calcium phosphate micron-sized rods. Microporous and Mesoporous Materials. 2006, 94 (1-3), 330-338.