Thermo Scientific Chemicals

mono-n-Dodecyl phosphate, tech. 90%, Thermo Scientific Chemicals

Catalog number: L11941.14
25 g, Each
Thermo Scientific Chemicals

mono-n-Dodecyl phosphate, tech. 90%, Thermo Scientific Chemicals

Catalog number: L11941.14
25 g, Each
Quantity
Catalog number: L11941.14
also known as L11941-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2627-35-2
Assay (Silylated GC)
≥88.0%
Assay (Aqueous acid-base Titration)
≥88.0%
Appearance (Color)
White
Form
Crystals or powder or crystalline powder

Description

mono-n-Dodecyl phosphate is a lysophosphatidic acid (LPA) receptor ligand which serves as a specific agonist at EDG-4 (LPA2) (EC50=700 nM) and antagonist at EDG-7

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
mono-n-Dodecyl phosphate is a lysophosphatidic acid (LPA) receptor ligand which serves as a specific agonist at EDG-4 (LPA2) (EC50=700 nM) and antagonist at EDG-7

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text