Thermo Scientific Chemicals

2-Bromobenzyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L11953.06
5 g, Each
Thermo Scientific Chemicals

2-Bromobenzyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L11953.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L11953.06
also known as L11953-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
578-51-8
IUPAC Name
1-bromo-2-(chloromethyl)benzene
Molecular Formula
C7H6BrCl
InChI Key
DDVSFIUKWUTKES-UHFFFAOYSA-N
SMILES
ClCC1=CC=CC=C1Br
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥96.0%
Form
Liquid
Refractive Index
1.5835-1.5885 @ 20°C

Description

2-Bromobenzyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text