3,5-Di-tert-butyltoluene, 98+%, Thermo Scientific Chemicals
3,5-Di-tert-butyltoluene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Di-tert-butyltoluene, 98+%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Catalog number L11961.14
also known as L11961-14
Price (USD)/ Each
67.65
Online exclusive
74.60 
Save 6.95 (9%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
67.65
Online exclusive
74.60 
Save 6.95 (9%)
Add to cart
3,5-Di-tert-butyltoluene, 98+%, Thermo Scientific Chemicals
Catalog numberL11961.14
Price (USD)/ Each
67.65
Online exclusive
74.60 
Save 6.95 (9%)
-
Add to cart
Chemical Identifiers
CAS15181-11-0
IUPAC Name1,3-di-tert-butyl-5-methylbenzene
Molecular FormulaC15H24
InChI KeyWIXDSJRJFDWTNY-UHFFFAOYSA-N
SMILESCC1=CC(=CC(=C1)C(C)(C)C)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White or clear colorless
FormFused solid or chunks or liquid as melt
Assay (GC)≥98.0%
Refractive Index1.4880-1.4920 as melt
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only