Thermo Scientific Chemicals

2,5-Dichlorobenzylamine, 97%, Thermo Scientific Chemicals

Catalog number: L11972.06
5 g, Each
Thermo Scientific Chemicals

2,5-Dichlorobenzylamine, 97%, Thermo Scientific Chemicals

Catalog number: L11972.06
5 g, Each
Quantity
Catalog number: L11972.06
also known as L11972-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10541-69-2
IUPAC Name
1-(2,5-dichlorophenyl)methanamine
Molecular Formula
C7H7Cl2N
InChI Key
AKGJLIXNRPNPCH-UHFFFAOYSA-N
SMILES
NCC1=CC(Cl)=CC=C1Cl
Assay (GC)
≥96.0%
Refractive Index
1.5750-1.5800 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid

Description

2,5-Dichlorobenzylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dichlorobenzylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Not miscible or difficult to mix.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Handle and store under inert gas. Incompatible with strong oxidizing agents. It is sensitive to air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text