Thermo Scientific Chemicals

Cyclopropyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L11985.06
5 g, Each
Thermo Scientific Chemicals

Cyclopropyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L11985.06
5 g, Each
Quantity
Catalog number: L11985.06
also known as L11985-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56601-42-4
IUPAC Name
isothiocyanatocyclopropane
Molecular Formula
C4H5NS
InChI Key
JGFBQFKZKSSODQ-UHFFFAOYSA-N
SMILES
S=C=NC1CC1
Assay (GC)
≥96.0% (non-U.S. specification)
Assay from Suppliers CofA
≥96.0% (U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated
Refractive Index
1.5465-1.5525 @ 20°C (non-U.S. specification)
Appearance (Color)
Clear colorless to pale yellow or pale brown

Description

Cyclopropyl isothiocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropyl isothiocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text