Thermo Scientific Chemicals

3-(Trifluoromethyl)phenylhydrazine, 95%, Thermo Scientific Chemicals

Catalog number: L11996.03
1 g, Each
Thermo Scientific Chemicals

3-(Trifluoromethyl)phenylhydrazine, 95%, Thermo Scientific Chemicals

Catalog number: L11996.03
1 g, Each
Quantity
Catalog number: L11996.03
also known as L11996-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
368-78-5
IUPAC Name
[3-(trifluoromethyl)phenyl]hydrazine
Molecular Formula
C7H7F3N2
InChI Key
RSESUCWJKLHXEZ-UHFFFAOYSA-N
SMILES
NNC1=CC=CC(=C1)C(F)(F)F
Assay (GC)
>94.0%
Refractive Index
1.5010-1.5070 @ 20?C
Appearance (Color)
Clear, colorless to yellow
Form
Liquid

Description

3-(Trifluoromethyl)phenylhydrazine is used in the synthesis of N-2-diphenylhydrazinecarboxamides and N-2-diphenylhydrazine carbothioamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Trifluoromethyl)phenylhydrazine is used in the synthesis of N-2-diphenylhydrazinecarboxamides and N-2-diphenylhydrazine carbothioamides.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text