3-Methylheptane, 97%, Thermo Scientific Chemicals
3-Methylheptane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methylheptane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number L12006.06
also known as L12006-06
Price (USD)/ Each
122.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
122.00
Add to cart
3-Methylheptane, 97%, Thermo Scientific Chemicals
Catalog numberL12006.06
Price (USD)/ Each
122.00
-
Add to cart
Chemical Identifiers
CAS589-81-1
IUPAC Name3-methylheptane
Molecular FormulaC8H18
InChI KeyLAIUFBWHERIJIH-UHFFFAOYNA-N
SMILESCCCCC(C)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.3955-1.4005 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only