Thermo Scientific Chemicals

3-(Methylthio)phenyl isocyanate, 96%, Thermo Scientific Chemicals

Catalog number: L12008.06
5 g, Each
Thermo Scientific Chemicals

3-(Methylthio)phenyl isocyanate, 96%, Thermo Scientific Chemicals

Catalog number: L12008.06
5 g, Each
Quantity
Catalog number: L12008.06
also known as L12008-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
28479-19-8
Identification (FTIR)
Conforms
Form
Liquid
Refractive Index
1.6000-1.6060 @ 20?C
Appearance (Color)
Clear colorless to yellow to green to pale brown
Assay (GC)
≥95.0%

Description

3-(Methylthio)phenyl isocyanate is used as a coupling reagent, N-protecting reagent and as gas chromatography derivatizing reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Methylthio)phenyl isocyanate is used as a coupling reagent, N-protecting reagent and as gas chromatography derivatizing reagent.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, amines and alcohols. It is sensitive to moisture. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text