Thermo Scientific Chemicals

4-Chloro-3-(trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L12016.06
5 g, Each
Thermo Scientific Chemicals

4-Chloro-3-(trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L12016.06
5 g, Each
Quantity
Catalog number: L12016.06
also known as L12016-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
327-78-6
IUPAC Name
1-chloro-4-isocyanato-2-(trifluoromethyl)benzene
Molecular Formula
C8H3ClF3NO
InChI Key
NBJZEUQTGLSUOB-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC(=CC=C1Cl)N=C=O
Appearance (Color)
Colorless to white
Form
Crystals or powder or crystalline powder or fused solid or clear liquid as melt
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text