Thermo Scientific Chemicals

4-Nitrophenyl octanoate, 96%, Thermo Scientific Chemicals

Catalog number: L12022.03
1 g, Each
Thermo Scientific Chemicals

4-Nitrophenyl octanoate, 96%, Thermo Scientific Chemicals

Catalog number: L12022.03
1 g, Each
Quantity
Catalog number: L12022.03
also known as L12022-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1956-10-1
IUPAC Name
4-nitrophenyl octanoate
Molecular Formula
C14H19NO4
InChI Key
GGIDEJQGAZSTES-UHFFFAOYSA-N
SMILES
CCCCCCCC(=O)OC1=CC=C(C=C1)[N+]([O-])=O
Identification (FTIR)
Conforms
Appearance (Color)
Clear pale yellow to yellow to brown
Assay (GC)
≥95.0%
Refractive Index
1.5075-1.5155 @ 20?C
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text