Thermo Scientific Chemicals

1-Adamantaneacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L12029.03
1 g, Each
Thermo Scientific Chemicals

1-Adamantaneacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L12029.03
1 g, Each
Quantity
Catalog number: L12029.03
also known as L12029-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4942-47-6
Melting Point (clear melt)
133.0-139.0?C
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥98.0%
Appearance (Color)
White to pale cream
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

1-Adamantaneacetic acid is used as an acylating agent in determining pharmacological characteristics of ten new analogues of bradykinin (Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg) that were modified in the N-terminal part of the molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantaneacetic acid is used as an acylating agent in determining pharmacological characteristics of ten new analogues of bradykinin (Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg) that were modified in the N-terminal part of the molecule.

Solubility
Solubility in Methanol - almost transparency

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text