2-(4-Morpholinyl)ethyl isothiocyanate is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(4-Morpholinyl)ethyl isothiocyanate is used as pharmaceutical intermediates.
Solubility
Hydrolyzes in water.
Notes
Store at 4°C. Store under dry inert gas. Protect from humidity and water. Incompatible with moisture, amines, alcohols and oxidizing agents. Protect from heat. Moisture Sensitive.
RUO – Research Use Only
Claude Guillonneau; Yves Charton; Yves-Michel Ginot; Marie-Victoire Fouquier-d’Hérouël; Marc Bertrand; Brian Lockhart; Pierre Lestage; Solo Goldstein. Synthesis and pharmacological evaluation of new 1,2-dithiolane based antioxidants. European Journal of Medicinal Chemistry.2003, 38, 1-11.