Thermo Scientific Chemicals

5-Hydroxy-2-nitrobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: L12051.06
5 g, Each
Thermo Scientific Chemicals

5-Hydroxy-2-nitrobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: L12051.06
5 g, Each
Quantity
Catalog number: L12051.06
also known as L12051-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
42454-06-8
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
163.0-173.0?C
Assay (GC)
≥96.0%
Appearance (Color)
Pale yellow to yellow to brown

Description

5-Hydroxy-2-nitrobenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Hydroxy-2-nitrobenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with air, oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text