Thermo Scientific Chemicals

1-(4-Methoxyphenyl)ethanol, 95%, Thermo Scientific Chemicals

Catalog number: L12062.22
100 g, Each
Thermo Scientific Chemicals

1-(4-Methoxyphenyl)ethanol, 95%, Thermo Scientific Chemicals

Catalog number: L12062.22
100 g, Each
Quantity
Catalog number: L12062.22
also known as L12062-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3319-15-1
IUPAC Name
1-(4-methoxyphenyl)ethan-1-ol
Molecular Formula
C9H12O2
InChI Key
IUUULXXWNYKJSL-UHFFFAOYNA-N
SMILES
COC1=CC=C(C=C1)C(C)O
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥94.0%
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.5300-1.5360 @ 20?C

Description

1-(4-Methoxyphenyl)ethanol is used to study the steady-state and nanosecond, laser-flash photolysis. It is used to produce 4-(1-chloro-ethyl)-anisole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(4-Methoxyphenyl)ethanol is used to study the steady-state and nanosecond, laser-flash photolysis. It is used to produce 4-(1-chloro-ethyl)-anisole.

Solubility
Not miscible in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text