This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Not miscible in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
RUO – Research Use Only
Hidetaka Nagatomo; Yoh-ichi Matsushita; Kazuhiro Sugamoto; Takanao Matsui. Preparation and properties of gelatin-immobilized beta-glucosidase from Pyrococcus furiosus. Bioscience, Biotechnology, and Biochemistry.2005, 69, (1), 128-136.