Bis(2-benzamidophenyl) disulfide, 97%, Thermo Scientific Chemicals
Bis(2-benzamidophenyl) disulfide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(2-benzamidophenyl) disulfide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number L12110.22
also known as L12110-22
Price (USD)/ Each
95.65
Online exclusive
106.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
95.65
Online exclusive
106.00 
Save 10.35 (10%)
Add to cart
Bis(2-benzamidophenyl) disulfide, 97%, Thermo Scientific Chemicals
Catalog numberL12110.22
Price (USD)/ Each
95.65
Online exclusive
106.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS135-57-9
IUPAC NameN-{2-[(2-benzamidophenyl)disulfanyl]phenyl}benzamide
Molecular FormulaC26H20N2O2S2
InChI KeyZHMIOPLMFZVSHY-UHFFFAOYSA-N
SMILESO=C(NC1=CC=CC=C1SSC1=CC=CC=C1NC(=O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)137.0-147.0?C
Appearance (Color)Yellow to green
Assay (HPLC)≥96.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only