Thermo Scientific Chemicals

1,1,1,3,3,3-Hexafluoroisopropyl acrylate, 98+%, stab. with 50ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L12128.14
25 g, Each
Thermo Scientific Chemicals

1,1,1,3,3,3-Hexafluoroisopropyl acrylate, 98+%, stab. with 50ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L12128.14
25 g, Each
Quantity
Catalog number: L12128.14
also known as L12128-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2160-89-6
IUPAC Name
1,1,1,3,3,3-hexafluoropropan-2-yl prop-2-enoate
Molecular Formula
C6H4F6O2
InChI Key
MNSWITGNWZSAMC-UHFFFAOYSA-N
SMILES
FC(F)(F)C(OC(=O)C=C)C(F)(F)F
Form
Liquid
Appearance (Color)
Clear colorless
Refractive Index
1.3170-1.3210 @ 20°C
Assay (GC)
≥97.5% (main component)
Solution Test
(10% v/v in methanol) : Clear

Description

1,1,1,3,3,3-Hexafluoroisopropyl acrylate is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text