Thermo Scientific Chemicals

Dichlorodimethylsilane, 98+%, Thermo Scientific Chemicals

Catalog number: L12133.22
100 g, Each
Thermo Scientific Chemicals

Dichlorodimethylsilane, 98+%, Thermo Scientific Chemicals

Catalog number: L12133.22
100 g, Each
Quantity
Catalog number: L12133.22
also known as L12133-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-78-5
IUPAC Name
dichlorodimethylsilane
Molecular Formula
C2H6Cl2Si
InChI Key
LIKFHECYJZWXFJ-UHFFFAOYSA-N
SMILES
C[Si](C)(Cl)Cl
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4025-1.4065 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥98.0%

Description

Dichlorodimethylsilane is used to prepare a resin bound siloxane with tertiary alcohols and it is also used as a reagent for synthesis of optically active ansa-mettallocene polymerization catalysts. It acts as a precursor to silicone and polysilane compounds. It is also used in the glass coating to protect it from micro particles. It is involved in the preparation of resin bound siloxane with reactivity towards tertiary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichlorodimethylsilane is used to prepare a resin bound siloxane with tertiary alcohols and it is also used as a reagent for synthesis of optically active ansa-mettallocene polymerization catalysts. It acts as a precursor to silicone and polysilane compounds. It is also used in the glass coating to protect it from micro particles. It is involved in the preparation of resin bound siloxane with reactivity towards tertiary alcohols.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with alcohols, amines and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text