(1-Decyl)trimethylammonium bromide is used in high performance liquid chromatography and in nuclear magnetic resonance. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(1-Decyl)trimethylammonium bromide is used in high performance liquid chromatography and in nuclear magnetic resonance. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. It is hygroscopic in nature. Incompatible with strong oxidizing agents.
RUO – Research Use Only