Thermo Scientific Chemicals

2-Methoxy-4-nitrophenyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L12150.03
1 g, Each
Thermo Scientific Chemicals

2-Methoxy-4-nitrophenyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L12150.03
1 g, Each
Quantity
Catalog number: L12150.03
also known as L12150-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
190774-55-1
IUPAC Name
1-isothiocyanato-2-methoxy-4-nitrobenzene
Molecular Formula
C8H6N2O3S
InChI Key
NXWXXLFRMVILJN-UHFFFAOYSA-N
SMILES
COC1=C(C=CC(=C1)[N+]([O-])=O)N=C=S
Appearance (Color)
Yellow to green to red to brown
Form
Powder or lumps
Melting Point (clear melt)
105.0-115.0?C
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text