3-(Trifluoromethyl)butyric acid, 97%, Thermo Scientific Chemicals
3-(Trifluoromethyl)butyric acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(Trifluoromethyl)butyric acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L12160.06
also known as L12160-06
Price (USD)/ Each
419.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
419.00
Add to cart
3-(Trifluoromethyl)butyric acid, 97%, Thermo Scientific Chemicals
Catalog numberL12160.06
Price (USD)/ Each
419.00
-
Add to cart
Chemical Identifiers
CAS348-75-4
IUPAC Name4,4,4-trifluoro-3-methylbutanoic acid
Molecular FormulaC5H7F3O2
InChI KeyFFZMMBKGTNDVRX-UHFFFAOYNA-N
SMILESCC(CC(O)=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (GC)≥96.0% (non-U.S. specification)
Refractive Index1.3565-1.3615 @ 20°C (non-U.S. specification)
FormLiquid
Identification (FTIR)Conforms (U.S. specification)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only