o-Tolyl isocyanate is used as plastic additives and as heat treatment salt formulations for metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
o-Tolyl isocyanate is used as plastic additives and as heat treatment salt formulations for metals.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Incompatible with bases, moisture and oxidizing agents. Store under dry inert gas. Protect from humidity and water.
RUO – Research Use Only
Hans J. Hess; Timothy H. Cronin; Alexander Scriabine. Antihypertensive 2-amino-4(3H)-quinazolinones. J. Med. Chem.1968, 11, (1), 130-136.
I.C Kogon. Chemistry of Aryl Isocyanates: Rate and Equilibrium Constants for the Formation of Ethyl α,γ-Diarylallophanate. J. Org. Chem.1959, 24, (1), 83-86.