Thermo Scientific Chemicals

Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals

Catalog number: L12265.18
50 g, Each
Thermo Scientific Chemicals

Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals

Catalog number: L12265.18
50 g, Each
Quantity
Catalog number: L12265.18
also known as L12265-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58052-80-5
IUPAC Name
2,4,6-trimethyl-1,3,5-triazinane-1,3,5-triium
Molecular Formula
C6H18N3
InChI Key
MZSSRMMSFLVKPK-UHFFFAOYSA-Q
SMILES
CC1[NH2+]C(C)[NH2+]C(C)[NH2+]1
Form
Crystals or powder or crystalline powder or fused solid
Assay from Supplier's CofA
≥97.5% (US-sourced material)
Identification (FTIR)
Conforms (UK-sourced material)
Appearance (Color)
White to pale cream to cream to yellow
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5% (UK-sourced material)

Description

Acetaldehyde ammonia trimer is used as scavengers for sulfhydryl compounds in natural gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetaldehyde ammonia trimer is used as scavengers for sulfhydryl compounds in natural gas.

Solubility
Soluble in water.

Notes
Desiccate at 4°C. Store away from air, moisture and oxidizing agents
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text